missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC17A4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | SLC17A4 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18285032
|
Novus Biologicals
NBP2-57090 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18638638
|
Novus Biologicals
NBP2-57090-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC17A4 Polyclonal specifically detects SLC17A4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SLC17A4 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| KAIA2138, KIAA2138, MGC129623, Na/PO4 cotransporter, putative small intestine sodium-dependent phosphate transport protein, solute carrier family 17 (sodium phosphate), member 4, Solute carrier family 17 member 4 | |
| SLC17A4 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 10050 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:YDDPVNHPFISAGEKRYIVCSLAQQDCSPGWSLPIRA | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title