missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC17A1/NPT Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£174.00 - £364.00
Specifications
| Antigen | SLC17A1/NPT |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18678012
|
Novus Biologicals
NBP2-94745-0.02ml |
0.02 mL |
£174.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610272
|
Novus Biologicals
NBP2-94745-0.1ml |
0.1 mL | |||||||
Description
SLC17A1/NPT Polyclonal antibody specifically detects SLC17A1/NPT in Rat samples. It is validated for Western BlotSpecifications
| SLC17A1/NPT | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat | |
| sodium-dependent phosphate transport protein 1, NAPI-1, NPT1, NPT-1, solute carrier family 17 (sodium phosphate), member 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SLC17A1 (NP_005065.2). MQMDNRLPPKKVPGFCSFRYGLSFLVHCCNVIITAQRACLNLTMVVMVNSTDPHGLPNTSTKKLLDNIKNPMYNWSPDIQGIILSSTSYGVIIIQVPVGY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 6568 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title