missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC17A1/NPT Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94745-0.1ml
This item is not returnable.
View return policy
Description
SLC17A1/NPT Polyclonal antibody specifically detects SLC17A1/NPT in Rat samples. It is validated for Western Blot
Specifications
| SLC17A1/NPT | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| sodium-dependent phosphate transport protein 1, NAPI-1, NPT1, NPT-1, solute carrier family 17 (sodium phosphate), member 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SLC17A1 (NP_005065.2). MQMDNRLPPKKVPGFCSFRYGLSFLVHCCNVIITAQRACLNLTMVVMVNSTDPHGLPNTSTKKLLDNIKNPMYNWSPDIQGIILSSTSYGVIIIQVPVGY | |
| 0.1 mL | |
| Primary | |
| Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6568 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction