missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC12A3 Monoclonal Antibody (C2)
GREENER_CHOICE

Mouse Monoclonal Antibody

£177.00 - £458.00

Specifications

Antigen SLC12A3
Clone C2
Concentration 1 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17827602
GREENER_CHOICE
View Documents
Invitrogen™
MA541643
200 μL
£458.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17694026
View Documents
Invitrogen™
MA543573
20 μL
£177.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SLC12A3 Monoclonal Antibody for Western Blot

The SLC12A3 gene encodes the thiazide-sensitive sodium-chloride symporter (NCC), which is critical for salt balance and blood pressure regulation in humans. This symporter is predominantly expressed in the distal convoluted tubule of the kidney, where it facilitates the reabsorption of sodium and chloride ions from the renal filtrate back into the bloodstream. NCC activity plays a vital role in maintaining extracellular fluid volume and potassium homeostasis, impacting systemic blood pressure. Mutations in SLC12A3 can lead to Gitelman syndrome, a hereditary salt-wasting disorder characterized by hypokalemic metabolic alkalosis, low blood pressure, and hypomagnesemia. This gene is regulated by the renin-angiotensin system, which modulates NCC activity through phosphorylation mediated by WNK kinases, thereby affecting renal sodium handling and blood pressure modulation. Research highlights the therapeutic potential of targeting NCC with thiazide diuretics to treat hypertension, emphasizing its importance in both physiological and pathological contexts. The dynamic regulation of NCC by hormonal pathways underscores its significance in fluid and electrolyte balance, contributing to cardiovascular health.
Specifications

Specifications

SLC12A3
1 mg/mL
Western Blot
Unconjugated
Mouse
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P55017
110260927, 6559
Recombinant protein Na-Cl Cotransporter. The antigen corresponds to amino acid range 3-146 of the target protein.
Antibody
C2
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Monoclonal
Liquid
IgG2b κ
Human, Pig
Slc12a3
AI035291; chemokine (C-C); CKb10; CK-beta-10; MCP-4; MGC17134; Na-Cl cotransporter; NaCl electroneutral thiazide-sensitive cotransporter; Na-Cl symporter; Ncc; NCC1; NCC-1; NCCT; SCYA13; SCYL1; si:ch211-122c9.4; SLC12A3; sodium chloride cotransporter; solute carrier family 12 (sodium/chloride transporter), member 3; solute carrier family 12 (sodium/chloride transporters), member 3; solute carrier family 12 member 3; solute carrier family 12, member 3; thiazide-sensitive Na-Cl cotransporter; thiazide-sensitive sodium chloride cotransporter; thiazide-sensitive sodium-chloride cotransporter; TSC; wu:fj83c03
Slc12a3
Primary
Protein A/G
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.