missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC12A3 Monoclonal Antibody (C2)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543573
This item is not returnable.
View return policy
Description
SLC12A3 Monoclonal Antibody for Western Blot
The SLC12A3 gene encodes the thiazide-sensitive sodium-chloride symporter (NCC), which is critical for salt balance and blood pressure regulation in humans. This symporter is predominantly expressed in the distal convoluted tubule of the kidney, where it facilitates the reabsorption of sodium and chloride ions from the renal filtrate back into the bloodstream. NCC activity plays a vital role in maintaining extracellular fluid volume and potassium homeostasis, impacting systemic blood pressure. Mutations in SLC12A3 can lead to Gitelman syndrome, a hereditary salt-wasting disorder characterized by hypokalemic metabolic alkalosis, low blood pressure, and hypomagnesemia. This gene is regulated by the renin-angiotensin system, which modulates NCC activity through phosphorylation mediated by WNK kinases, thereby affecting renal sodium handling and blood pressure modulation. Research highlights the therapeutic potential of targeting NCC with thiazide diuretics to treat hypertension, emphasizing its importance in both physiological and pathological contexts. The dynamic regulation of NCC by hormonal pathways underscores its significance in fluid and electrolyte balance, contributing to cardiovascular health.
Specifications
| SLC12A3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P55017 | |
| Slc12a3 | |
| Recombinant protein Na-Cl Cotransporter. The antigen corresponds to amino acid range 3-146 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG2b κ |
| Western Blot | |
| C2 | |
| Unconjugated | |
| Slc12a3 | |
| AI035291; chemokine (C-C); CKb10; CK-beta-10; MCP-4; MGC17134; Na-Cl cotransporter; NaCl electroneutral thiazide-sensitive cotransporter; Na-Cl symporter; Ncc; NCC1; NCC-1; NCCT; SCYA13; SCYL1; si:ch211-122c9.4; SLC12A3; sodium chloride cotransporter; solute carrier family 12 (sodium/chloride transporter), member 3; solute carrier family 12 (sodium/chloride transporters), member 3; solute carrier family 12 member 3; solute carrier family 12, member 3; thiazide-sensitive Na-Cl cotransporter; thiazide-sensitive sodium chloride cotransporter; thiazide-sensitive sodium-chloride cotransporter; TSC; wu:fj83c03 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 110260927, 6559 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction