missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC10A7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£388.00
Specifications
| Antigen | SLC10A7 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SLC10A7 Polyclonal specifically detects SLC10A7 in Human samples. It is validated for Western Blot.Specifications
| SLC10A7 | |
| Polyclonal | |
| Rabbit | |
| Q0GE19-2 | |
| 84068 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| C4orf13, chromosome 4 open reading frame 13, DKFZp313H0531, DKFZp566M114, DKFZp779O2438, MGC25043, Na(+)/bile acid cotransporter 7, P7, SBF-domain containing protein, sodium/bile acid cotransporter 7, solute carrier family 10 (sodium/bile acid cotransporter family), member 7, Solute carrier family 10 member 7 | |
| SLC10A7 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title