missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slanetz and Bartley Medium

Product Code. 12769090
Change view
Click to view available options
Format:
55 mm monoplate
90 mm Monoplate
Fill Volume:
15 mL
17 mL
Fill Weight:
13 g ± 5%
17g ± 5%
Quantity:
10/Pk.
2 x 1 x 10
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Fill Volume Fill Weight Quantity Unit Size
12769090 90 mm Monoplate 17 mL 17g ± 5% 10/Pk. Pack of 10
11998419 55 mm monoplate 15 mL 13 g ± 5% 2 x 1 x 10 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12769090 Supplier Thermo Scientific™ Supplier No. PO5018A
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect and enumerate enterococci from water and food samples with Thermo Scientific™ Slanetz and Bartley Medium.

Detect and enumerate enterococci from water and food samples with Thermo Scientific™ Slanetz and Bartley Medium. Slanetz and Bartley1 originally devised this medium to detect and enumerate enterococci by the technique of membrane filtration, but it has also proved useful as a direct plating medium2,3. The medium is very selective for enterococci, and when it is incubated at elevated temperatures (44-45°C), all red or maroon colonies may be accepted as presumptive enterococci4,5. The medium has been tested in accordance with the methods described in ISO 7704:2023.

The Environment Agency Microbiology of Drinking Water 20026 recommend the use of Slanetz and Bartley medium for the enumeration of enterococci in water supplies, as does ISO in the standard for water quality7.

The water is filtered through a membrane filter which is then placed on the surface of a well dried plate of the medium. Plates are incubated at 35°C for 4 hours and then at 44-45°C for 44 hours. Membranes are examined, with a hand lens in a good light, and all red or maroon colonies counted as enterococci.

Food samples can be examined for enterococci by the method suggested by the Nordic Committee of Food Analysis3. Samples are homogenizsed and so diluted with physiological saline that only 15 to 150 colonies grow on each Petri dish. Homogenates or dilutions are spread evenly over the agar surface with a glass rod and allowed to soak in. Dishes are inverted and incubated at 35°C for 48 hours, after which typical colonies (pink or dark red, with a narrow whitish border) are counted.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Beige to beige red, transparent
Content And Storage 2° to 12°C, in the dark
Description Slanetz and Bartley Medium
Format 90 mm Monoplate
Manufacturing Origin Germany
Product Type Microbiology Media
Fill Volume 17 mL
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.