missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Skp1 Rabbit anti-Human, Mouse, Rat, Clone: 10G5X5, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £395.00
Specifications
| Antigen | Skp1 |
|---|---|
| Clone | 10G5X5 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18311327
|
Novus Biologicals
NBP3-16734-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18302227
|
Novus Biologicals
NBP3-16734-100UL |
100 μg |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Skp1 Monoclonal antibody specifically detects Skp1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Skp1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Wnt Signaling Pathway | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 6500 | |
| IgG | |
| Affinity purified |
| 10G5X5 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Cyclin-A/CDK2-associated protein p19, EMC19p19skp1, MGC34403, OCP2OCP-2, OCP-IIS-phase kinase-associated protein 1A (p19A), Organ of Corti protein 2, Organ of Corti protein II, p19Acyclin A/CDK2-associated p19, RNA polymerase II elongation factor-like protein, RNA polymerase II elongation factor-like protein OCP2, SKP1Acyclin A/CDK2-associated protein p19, S-phase kinase-associated protein 1, TCEB1LSIII, Transcription elongation factor B | |
| A synthetic peptide corresponding to a sequence within amino acids 64-163 of human Skp1 (P63208). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title