missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Skp1 Rabbit anti-Human, Mouse, Rat, Clone: 10G5X5, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16734-100UL
This item is not returnable.
View return policy
Description
Skp1 Monoclonal antibody specifically detects Skp1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Skp1 | |
| Monoclonal | |
| Unconjugated | |
| Cyclin-A/CDK2-associated protein p19, EMC19p19skp1, MGC34403, OCP2OCP-2, OCP-IIS-phase kinase-associated protein 1A (p19A), Organ of Corti protein 2, Organ of Corti protein II, p19Acyclin A/CDK2-associated p19, RNA polymerase II elongation factor-like protein, RNA polymerase II elongation factor-like protein OCP2, SKP1Acyclin A/CDK2-associated protein p19, S-phase kinase-associated protein 1, TCEB1LSIII, Transcription elongation factor B | |
| A synthetic peptide corresponding to a sequence within amino acids 64-163 of human Skp1 (P63208). | |
| 100 μg | |
| Wnt Signaling Pathway | |
| 6500 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| 10G5X5 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction