missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIX5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22938-1-AP-150UL
This item is not returnable.
View return policy
Description
Six5 (homeobox protein SIX5), also known as SIX5, BOR2 or DMAHP (DM locus-associated homeodomain protein), is a transcription factor that is expressed in various structures of the adult eye. Localized to the cytoplasm in early development and to the nucleus in the later stages of development,Six5 is involved in regulation of organogenesis and in maintece of retinal formation. Six5 is able to bind the 5′-TCA[AG][AG]TTNC-3′ DNA sequence found in the myogenin and IGFBP5 promoters and, through this binding, can control transcription of the associated mRNA. Six5 is regulated via association with DACH1 (dachshund homolog 1) and is co-activated by the EYA (eyes absent) proteins. Defects in the gene encoding Six5 are the cause of branchiooto- renal syndrome type 2 (BOR2), an autosomal disorder characterized by hearing loss, a deep overbite and myopia. Two isoforms exist due to alternative splicing events.Specifications
| SIX5 | |
| Polyclonal | |
| Unconjugated | |
| SIX5 | |
| BOR2, DMAHP, Homeobox protein SIX5, Sine oculis homeobox homolog 5, SIX homeobox 5, SIX5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 147912 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N196 | |
| SIX5 | |
| SIX5 Fusion Protein Ag18993 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction