missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SiteClick™ sDIBO Alkyne Kits for Antibody Labeling

Product Code. 16804076
Change view
Click to view available options
Label or Dye:
Alexa Fluor 488
Alexa Fluor 555
Alexa Fluor 647
Biotin
Primary Amine
pHrodo Deep Red
pHrodo Red
Labeling Scale:
1 mg
100-250 μg
5 mg
Unit Size:
1 set
1mg
5mg
Each
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Label or Dye Labeling Scale Unit Size
16804076 Alexa Fluor 488 1 mg 1mg
15980573 Alexa Fluor 488 100-250 μg Each
15998810 Alexa Fluor 647 100-250 μg Each
15910583 Primary Amine 100-250 μg Each
15956065 pHrodo Red 100-250 μg 1 set
17133783 Biotin 1 mg 1mg
17143783 pHrodo Red 1 mg 1mg
17153783 Alexa Fluor 647 1 mg 1mg
17163783 Biotin 5 mg 5mg
17173783 pHrodo Red 5 mg 1mg
17183783 Alexa Fluor 488 5 mg 1mg
17193783 Alexa Fluor 647 5 mg 1mg
17172534 pHrodo Deep Red 100-250 μg 1 set
17182534 pHrodo Deep Red 1 mg 1 set
15990573 Alexa Fluor 555 100-250 μg Each
15900583 Biotin 100-250 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Product Code. 16804076 Supplier Invitrogen™ Supplier No. S10904
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Easily attach Alexa Fluor™ pHrodo™ and Biotin sDIBO Alkyne tags to azido-modified antibodies using SiteClick™ chemistry.

Generate site-specific labeled antibody conjugates with SiteClick sDIBO Alkynes for Antibody Labeling. Our SiteClick sDIBO Antibody Labeling kits are optimized to be used in conjunction with the SiteClick Azido Modification kits (S20026, S10900, or S10901 sold separately) and contain sufficient reagents to perform a conjugation reaction of a SiteClick sDIBO Alkyne label using 100 μg, 1 mg, or 5 mg of azido-modified IgG antibody. The SiteClick sDIBO Alkynes for Antibody Labeling employs copper-free click chemistry to covalently link a dibenzocyclooctyne (sDIBO) labeled Alexa Fluor dye, pHrodo dye, or biotin to an azido modified antibody. The label, when attached to the antibody, is located far away from its antigen binding domain.

Create your own covalently labeled antibody conjugates using the SiteClick sDIBO Alkyne labels, which are intended for use with azido-modified antibodies. SiteClick sDIBO Alkyne Antibody Labeling Kits include the sDIBO Alkyne and the antibody concentrator, enabling copper-free covalent attachment of Alexa Fluor and pHrodo dyes, biotin, or molecules of your choice. When used with the SiteClick Antibody Azido Modification Kit (available separately as Cat. No. S20026, S10900, S10901), labeling occurs on the heavy chain N-linked glycans, far away from the antigen-binding domain, enabling excellent reproducibility between separate labeling reactions and multiple antibody species and subtypes. The SiteClick sDIBO alkyne kits for antibody labeling are optimized for conjugation reactions, covalently linking 100 μg, 1 mg, or 5 mg of azide-modified IgG antibody.

The SiteClick Antibody Azido Modification Kit (Cat. No. S20026, S10900, S10901) is recommended for the attachment of an azide moiety to the heavy chains of an unlabeled IgG antibody. Other engineered antibodies containing azido moieties are also suitable.

When used in conjunction with the SiteClick Antibody Azido Modification Kit, the SiteClick sDIBO alkyne kits for antibody labeling give you the option of trying different fluorescent labels for your antibody, attaching other molecules via streptavidin, or attaching your own molecule via amine-reactive moieties, depending on your assay. Our sDIBO alkynes are improved versions of DIBO cyclooctynes, yielding conjugates that are less 'sticky' and emit lower signal background in biological samples.

TRUSTED_SUSTAINABILITY

Specifications

Quantity for 1 mg of Ab
Kit Contents Contains 1 vial of SiteClick Alexa Fluor 488 sDIBO Alkyne for Antibody Labeling in 50 μL DMSO and 1 antibody concentrator
Filter Type GFP
Content And Storage Store in refrigerator (2°C to 8°C) and protect from light.
Chemical Reactivity Azide
Flow Cytometer Laser Lines Blue
Labeling Method Click Chemistry
Labeling Scale 1 mg
Shipping Condition Wet Ice
Reactive Moiety sDIBO Alkyne
Label or Dye Alexa Fluor 488
Color Green
Product Line Alexa Fluor, SiteClick
Labeling Target Azido modified antibodies
Excitation/Emission 490/525 nm
Show More Show Less
What is the general protocol for using the sDIBO alkynes?

The protocol is very simple. Expose the azide-modified sample to the sDIBO alkyne (final concentration in the range of 1 to 5 µM is a reasonable starting range) in aqueous buffer of media for 1-2 hrs. Optimize the final working concentration and incubation time per the sample/application. The azide-modified target must have the azide accessible for reacting with the sDIBO, and the labeling solution should not include any sodium azide.

How are the sDIBO alkynes different from the original DIBO alkynes?

Both the original DIBO alkynes and the sDIBO alkynes utilize the same fluorescent dyes or biotin for detection. The sDIBO alkynes have improved solubility in aqueous solutions and less propensity to bind non-specifically compared to the original DIBO alkynes.

How is SiteClick Alexa Fluor 488 sDIBO Alkyne different from Click-iT Alexa Fluor 488 sDIBO Alkyne?

Structurally, they are the same. The only difference is in the manner of packaging. SiteClick Alexa Fluor 488 sDIBO Alkyne is packaged for labeling 1 to 5 mg of antibody per the SiteClick labeling protocol with the actual amount being proprietary. Click-iT Alexa Fluor 488 sDIBO Alkyne is packaged as a 0.5 mg of reagent for use with any copper-free click reaction.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.