Learn More
Invitrogen™ SiteClick™ Duet and Quartet Antibody Labeling Kits
Description
SiteClick Duet and Quartet antibody labeling kits are used to create label-ready, site-specific, azido-modified antibodies without lengthy genetic modifications. Our enzyme-driven SiteClick labeling precisely attaches an azido moiety to the heavy chains of IgG antibodies, preserving antigen-binding domains for optimal target binding. This site selectivity targets carbohydrate domains on the Fc region, applicable to all IgG antibodies regardless of isotype or host species.
The SiteClick Quartet Antibody Labeling Kit offers up to four payloads per antibody (DOS 4) in less than 24 hours, while the SiteClick Duet Antibody Labeling Kit enables the attachment of two payloads (DOS 2) in only 4 hours.
Features of the SiteClick Duet and Quartet antibody labeling kits include:
- Modify any IgG antibody species or isotype with a complete kit
- Follow a brief, straightforward protocol
- Preserve antibody function with efficient, site-specific labeling
- Achieve consistent results with reproducible labeling chemistry
- Develop therapeutic antibodies, antibody-drug conjugates, antibody-oligo conjugates, bispecific antibodies, and immunodiagnostics
Once azido-modified, choose from a variety of SiteClick sDIBO or DBCO alkyne labels for covalent attachment via copper-free click chemistry under mild conditions. Enjoy the flexibility to select different fluorescent labels, including Alexa Fluor, pHrodo, and LysoLight dyes, or to conjugate other molecules such as biotin, oligos, toxins, or degraders.
SiteClick labeling is useful for developing therapeutic antibodies, antibody-drug conjugates, biologics, and immunodiagnostic tests, helping ensure high specificity, reproducible labeling, and preserved antibody performance.
Custom SiteClick Antibody Labeling Service
Unlock the full potential of your antibodies with our Custom SiteClick Antibody Labeling Service and customized labels. If conventional methods have failed or reduced your antibody's activity, our expert team can help. We offer SiteClick-compatible labels in bulk format, and custom labeling with a variety of detection molecules, including Alexa Fluor, LysoLight, and pHrodo dyes, as well as biotin and R-PE.
Interested in using SiteClick technology beyond your internal research? Let us help you use this in your product manufacturing and services. Please inquire about licensing options for your commercial applications.
Specifications
Specifications
| Green Features | Improved stability, less waste |
| For Use With (Application) | Antibody Production and Labeling |
| Product Type | Quartet Antibody Modification Kit |
| Quantity | 5 mg |
| Sufficient For | 1 Labeling |
| Content And Storage | 2°C to 8°C |
| For Use With (Equipment) | Benchtop Centrifuge |
| Labeling Method | Click Chemistry |
| Shipping Condition | Wet Ice |
| Product Line | SiteClick |
| Show More |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.