missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sirtuin 2/SIRT2, Mouse anti-Human, Clone: S2R233-1, Novus Biologicals™
Mouse Monoclonal Antibody
£280.00 - £376.00
Specifications
| Antigen | Sirtuin 2/SIRT2 |
|---|---|
| Clone | S2R233-1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18245326
|
Novus Biologicals
NBP2-80049-50UG |
50 μg |
£280.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18220078
|
Novus Biologicals
NBP2-80049-100UG |
100 μg |
£376.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sirtuin 2/SIRT2 Monoclonal specifically detects Sirtuin 2/SIRT2 in Human samples. It is validated for Western Blot, ELISA.Specifications
| Sirtuin 2/SIRT2 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Chromatin Research, DNA Repair, Epigenetics, Neuronal Cell Markers, Transcription Factors and Regulators | |
| 0.2 um-filtered solution in PBS, pH 7.4 | |
| 22933 | |
| Recombinant human Sirtuin 2/SIRT2. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S2R233-1 | |
| Western Blot, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.5.1, EC 3.5.1.-, FLJ35621, FLJ37491, S.cerevisiae, homolog) 2, sirtuin 2, sirtuin type 2 | |
| SIRT2 | |
| IgG2b κ | |
| Protein G purified | |
| Recognizes human Sirtuin 2/SIRT2. Detects a band of ∽45kDa by Western blot. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title