missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sirtuin 2/SIRT2, Mouse anti-Human, Clone: S2R233-1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80049-100UG
This item is not returnable.
View return policy
Description
Sirtuin 2/SIRT2 Monoclonal specifically detects Sirtuin 2/SIRT2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Sirtuin 2/SIRT2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| EC 3.5.1, EC 3.5.1.-, FLJ35621, FLJ37491, S.cerevisiae, homolog) 2, sirtuin 2, sirtuin type 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Recognizes human Sirtuin 2/SIRT2. Detects a band of ∽45kDa by Western blot. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA | |
| S2R233-1 | |
| Unconjugated | |
| 0.2 um-filtered solution in PBS, pH 7.4 | |
| SIRT2 | |
| Recombinant human Sirtuin 2/SIRT2. | |
| 100 μg | |
| Apoptosis, Cancer, Chromatin Research, DNA Repair, Epigenetics, Neuronal Cell Markers, Transcription Factors and Regulators | |
| 22933 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction