missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIRT6 Recombinant Rabbit Monoclonal Antibody (23GB1135), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | SIRT6 |
|---|---|
| Clone | 23GB1135 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30087675
|
Invitrogen™
MA553138 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088361
|
Invitrogen™
MA553139 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SIRT6 Recombinant Monoclonal antibody specifically detects SIRT6 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| SIRT6 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8N6T7 | |
| 51548 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1135 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2810449N18Rik; AI043036; NAD-dependent deacetylase sirtuin-6; NAD-dependent protein deacetylase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2; SIR2 like 6; Sir2l6; SIR2-like protein 6; sir2-related protein type 6; Sirt6; sirtuin 6; sirtuin type 6 | |
| A synthesized peptide derived from human SIRT6 (300-355AA). | |
| Primary | |
| -20°C | |
| SIRT6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title