missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIRT6 Recombinant Rabbit Monoclonal Antibody (23GB1135), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553138
This item is not returnable.
View return policy
Description
Sirtuins regulate epigenetic gene silencing. SIRT6 (Mono-ADP-ribosyltransferase sirtuin-6), a mouse Sir2 homolog, is a chromatin-associated nuclear ADP-ribosyltransferase protein. It appears to be involved in DNA repair and may also play a role in human aging.
Specifications
| SIRT6 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SIRT6 | |
| 2810449N18Rik; AI043036; NAD-dependent deacetylase sirtuin-6; NAD-dependent protein deacetylase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2; SIR2 like 6; Sir2l6; SIR2-like protein 6; sir2-related protein type 6; Sirt6; sirtuin 6; sirtuin type 6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51548 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1135 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8N6T7 | |
| SIRT6 | |
| A synthesized peptide derived from human SIRT6 (300-355AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction