missing translation for 'onlineSavingsMsg'
Learn More

SIRP beta 1/gamma, Rabbit anti-Human, Clone: OX117, Novus Biologicals™

Rabbit Monoclonal Antibody

£198.00 - £510.00

Specifications

Antigen SIRP beta 1/gamma
Clone OX117
Concentration 1 mg/mL
Dilution Flow Cytometry, ELISA, Immunoprecipitation
Applications Flow Cytometry, ELISA, Immunoprecipitation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18292349
View Documents
Novus Biologicals
NBP2-81072-0.025MG
0.025 mg
£198.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18379380
View Documents
Novus Biologicals
NBP2-81072-0.2MG
0.2 mg
£510.00
0.20mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SIRP beta 1/gamma Monoclonal specifically detects SIRP beta 1/gamma in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation.
Specifications

Specifications

SIRP beta 1/gamma
1 mg/mL
Flow Cytometry, ELISA, Immunoprecipitation
Unconjugated
Rabbit
Human
Recombinant human SIRPgamma:CD4 fusion protein.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
OX117
Flow Cytometry, ELISA, Immunoprecipitation
Monoclonal
Purified
RUO
PBS
IgG κ
Protein A purified
This antibody binds to human SIRP beta 1/gamma.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.