missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SIRP beta 1/gamma Monoclonal specifically detects SIRP beta 1/gamma in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation.
Specifications
Specifications
| Antigen | SIRP beta 1/gamma |
| Applications | Flow Cytometry, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | OX117 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry, ELISA, Immunoprecipitation |
| Formulation | PBS |
| Host Species | Rabbit |
| Immunogen | Recombinant human SIRPgamma:CD4 fusion protein. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?