Learn More
RETSCH Sintered Corundum Comfort Grinding Jars
Product Code. 10156541 Shop All Retsch Products
Change view
Click to view available options
Capacity (Metric):
50 mL
125 mL
250 mL
500 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10156541 125 mL Each
10348612 250 mL Each
10398032 500 mL Each
10207771 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10156541 Supplier RETSCH Supplier No. 01.462.0152
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these grinding jars with Retsch™ ball mills. Retsch™ Sintered Corundum Comfort Grinding Jars feature an O-type sealing for safe operation.
  • Available in a variety of sizes
  • Optional aeration covers
  • Pressure tight safety closure devices
Specifications
Product Type Comfort Grinding Jar
Capacity (Metric) 125 mL
For Use With (Equipment) For PM 100, PM 200, PM 400 and S 100
Material Sintered Corundum