Description
- Available in a variety of sizes
- Optional aeration covers
- Pressure tight safety closure devices
Specifications
Specifications
| Product Type | Comfort Grinding Jar |
| Capacity (Metric) | 250 mL |
| For Use With (Equipment) | For PM 100, PM 400 and S 100 |
| Material | Sintered Corundum |