missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF (TNFa) Recombinant Human Protein, Invitrogen™ Sino Biological™
This recombinant protein is expressed from a DNA sequence encoding the soluble form of human TNF (NP_000585.2) (Val 77-Leu 233) retaining the initial Met at the N-terminus
£57.25 - £452.00
Specifications
For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
---|---|
Molecular Weight (g/mol) | 17.4kDa |
Storage Requirements | Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
Structural Form | Full Length/Recombinant |
Gene Alias | TNF, DADB-70P7.1, DIF, TNF-alpha, TNFA, TNFSF2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13493019
|
Invitrogen
10602HNAE5 |
5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13473019
|
Invitrogen
10602HNAE25 |
5 x 5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13403029
|
Invitrogen
10602HNAE50 |
50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13483019
|
Invitrogen
10602HNAE250 |
5 x 50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Tumor necrosis factor alpha (TNF-α) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in analysis of protein structure and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.Background: Tumor necrosis factor alpha (TNF-α), also known as cachectin and TNFSF1A, is the prototypic cytokine of the TNF superfamily, and is a multifunctional molecule involved in the regulation of a wide spectrum of biological processes including cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF-α is produced mainly by macrophages, and large amounts of this cytokine are released in response to lipopolysaccharide, other bacterial products, and Interleukin-1 (IL-1). TNF-α is primarily produced as a type II transmembrane protein arranged in stable homotrimers, and a soluble trimeric form of TNF-α is released via proteolytic cleavage by the metalloprotease TACE/ ADAM17. Two receptors mediating the TNF-α function have been identified, the ubiquitous TNF RI from which most signaling are derived, and the hematopoietic cell-restricted TNF RII. Upon the TNF-α trimers binding, TNF receptors also form trimers, undergo a conformational change, and enable the adaptor protein TRADD to bind to the death domain, and thus initiates the signal pathways such as the NF-kB, Jak/STAT, and the MAPK pathways, as well as the death signaling. As a major mediator of apoptosis, inflammation and immunity, TNF-α has been implicated in the pathogenesis of a variety of human diseases including autoimmune diseases, insulin resistance, and cancer.
- N-terminal Sequence Analysis: Met
- Activity: measured in a cytotoxicity assay using L929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D, the EC50 for this effect is typically 25-200pg/mL, corresponding to a specific activity of 5 x 106 to 4 x 107 units/mg
- Formulation: lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% mannitol, 5% trehalose, 0.02% Tween™-80
- Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Manufactured by Sino Biological (China).
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
TNF, DADB-70P7.1, DIF, TNF-alpha, TNFA, TNFSF2 | |
TNF | |
E. coli | |
Lyophilized | |
≥95% by SDS-PAGE |
17.4kDa | |
Full Length/Recombinant | |
Tumor necrosis factor | |
Untagged | |
Cytokines & Receptors | |
TNF (Tumor Necrosis Factor) |
For Research Use Only. Not for use in diagnostic procedures.