Learn More
HA Recombinant Influenza A, Subtype H1N1 (A/Puerto Rico/8/1934) Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 528) of the influenza A hemagglutinin (A/Puerto Rico/8/1934 (H1N1)) (ABD77675.1), fused to a C-terminal polyhistidine tag
Brand: Invitrogen 11684V08H50
Additional Details : Weight : 0.01000kg
Description
Hemagglutinin (HA), recombinant viral protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.- Activity: This recombinant protein has not been tested
- Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
PAGE | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
Truncated/Recombinant | |
HA | |
HA | |
Mammalian | |
Lyophilized |
59kDa | |
50μg | |
C-Terminal | |
<1 EU/μg | |
Hemagglutinin | |
His Tag | |
Cell Structure/Organelle Proteins | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.