missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HA Recombinant Influenza A, Subtype H1N1 (A/Puerto Rico/8/1934) Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 528) of the influenza A hemagglutinin (A/Puerto Rico/8/1934 (H1N1)) (ABD77675.1), fused to a C-terminal polyhistidine tag
£210.00 - £662.00
Specifications
For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
---|---|
Molecular Weight (g/mol) | 59kDa |
Purification Method | PAGE |
Storage Requirements | Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
Termination | C-Terminal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15288845
|
Invitrogen
11684V08H50 |
50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15268845
|
Invitrogen
11684V08H250 |
5 x 50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Hemagglutinin (HA), recombinant viral protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.- Activity: This recombinant protein has not been tested
- Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
PAGE | |
C-Terminal | |
<1 EU/μg | |
Hemagglutinin | |
His Tag | |
Cell Structure/Organelle Proteins | |
≥90% by SDS-PAGE |
59kDa | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
Truncated/Recombinant | |
HA | |
HA | |
Mammalian | |
Lyophilized |
For Research Use Only. Not for use in diagnostic procedures.