Learn More
EGF Recombinant Mouse Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse EGF (P01132) (Asn 977-Arg 1029) fused to the Fc region of human IgG1 at the N-terminus
Brand: Invitrogen 50482M01H5
Code : NEW
Additional Details : Weight : 0.00001kg
Includes: One vial of lyophilized protein
Description
Epidermal growth factor (EGF), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.- Activity: Measured in a cell proliferation assay using BALB/c 3T3 mouse embryonic fibroblasts; the EC50 for this effect is typically 0.2-1ng/mL
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
5μg | |
Full Length/Recombinant | |
AI790464 | |
EGF | |
Mammalian | |
Lyophilized | |
≥90% by SDS-PAGE |
34.5kDa | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
Epidermal growth factor | |
IgG1 Fc Tag | |
Growth Factors & Receptors | |
EGF (Epidermal Growth Factor) |
For Research Use Only. Not for use in diagnostic procedures.