missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sin3A-associated protein Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £366.00
Specifications
| Antigen | Sin3A-associated protein |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18695372
|
Novus Biologicals
NBP2-94735-0.02ml |
0.02 mL |
£149.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18646932
|
Novus Biologicals
NBP2-94735-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sin3A-associated protein Polyclonal antibody specifically detects Sin3A-associated protein in Mouse samples. It is validated for Western BlotSpecifications
| Sin3A-associated protein | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| 130 kDa Sin3-associated polypeptide, histone deacetylase complex subunit SAP130, mSin3A-associated protein 130, SAP130, SAP130 Sin3A-associated protein, 130kDa, Sin3-associated polypeptide p130 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 819-1048 of human SAP130 (NP_078821.2). LSMPTSDLPPGASPRKKPRKQQHVISTEEGDMMETNSTDDEKSTAKSLLVKAEKRKSPPKEYIDEEGVRYVPVRPRPPITLLRHYRNPWKAAYHHFQRYSDVRVKEEKKAMLQEIANQKGVSCRAQGWKVHLCAAQLLQLTNLEHDVYERLTNLQEGIIPKKKAATDDDLHRINELIQGNMQRCKLVMDQISEARDSMLKVLDHKDRVLKLLNKNGTVKKVSKLKRKEKV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 79595 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title