missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sin3A-associated protein Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94735-0.1ml
This item is not returnable.
View return policy
Description
Sin3A-associated protein Polyclonal antibody specifically detects Sin3A-associated protein in Mouse samples. It is validated for Western Blot
Specifications
| Sin3A-associated protein | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 130 kDa Sin3-associated polypeptide, histone deacetylase complex subunit SAP130, mSin3A-associated protein 130, SAP130, SAP130 Sin3A-associated protein, 130kDa, Sin3-associated polypeptide p130 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 819-1048 of human SAP130 (NP_078821.2). LSMPTSDLPPGASPRKKPRKQQHVISTEEGDMMETNSTDDEKSTAKSLLVKAEKRKSPPKEYIDEEGVRYVPVRPRPPITLLRHYRNPWKAAYHHFQRYSDVRVKEEKKAMLQEIANQKGVSCRAQGWKVHLCAAQLLQLTNLEHDVYERLTNLQEGIIPKKKAATDDDLHRINELIQGNMQRCKLVMDQISEARDSMLKVLDHKDRVLKLLNKNGTVKKVSKLKRKEKV | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79595 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction