missing translation for 'onlineSavingsMsg'
Learn More

Silver perrhenate, 99.99% (metals basis)

Product Code. 11460820
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460820 5 g 5g
11450820 1 g 1g
11470820 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460820 Supplier Thermo Scientific Alfa Aesar Supplier No. 044480.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20654-56-2 | AgO4Re | 358.071 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20654-56-2
Molecular Formula AgO4Re
Molecular Weight (g/mol) 358.071
MDL Number MFCD00014144
InChI Key JRISYOCHUNKJAV-UHFFFAOYSA-N
Synonym silver i perrhenate, silver perrhenate, rhenium silver tetraoxide, acmc-20ajpz, silver i perrhenate, for synthesis, silver 1+ ion tetraoxorhenate 1-, silver i perrhenate trace metals basis
PubChem CID 11089628
IUPAC Name silver;oxido(trioxo)rhenium
SMILES [O-][Re](=O)(=O)=O.[Ag+]

Specifications

Melting Point 430°C
Color White
Physical Form Powder
Quantity 5 g
Linear Formula AgReO4
UN Number UN1479
Formula Weight 358.1
Percent Purity 99.99%
Sensitivity Light sensitive
Density 7.05 g/mL
Assay (metals basis)
Chemical Name or Material Silver perrhenate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.