CAS RN 20654-56-2
CAS RN 20654-56-2
IUPAC Name:
silver(1+) rheniumoylolate
Synonyms:
silver perrhenate
Molecular Weight (g/mol):
358.07
Molecular Formula:
AgO4Re
InChi Key:
JRISYOCHUNKJAV-UHFFFAOYSA-N
SMILES:
[Ag+].[O-][Re](=O)(=O)=O
1
–
1
of
1
results
1
Silver perrhenate, 99.99% (metals basis)
CAS: 20654-56-2 Molecular Formula: AgO4Re Molecular Weight (g/mol): 358.071 MDL Number: MFCD00014144 InChI Key: JRISYOCHUNKJAV-UHFFFAOYSA-N Synonym: silver i perrhenate,silver perrhenate,rhenium silver tetraoxide,acmc-20ajpz,silver i perrhenate, for synthesis,silver 1+ ion tetraoxorhenate 1-,silver i perrhenate trace metals basis PubChem CID: 11089628 IUPAC Name: silver;oxido(trioxo)rhenium SMILES: [O-][Re](=O)(=O)=O.[Ag+]