missing translation for 'onlineSavingsMsg'
Learn More

Silver hydrogen fluoride, 99% min, Thermo Scientific Chemicals

Product Code. 11369267
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369267 10 g 10g
11379267 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369267 Supplier Thermo Scientific Alfa Aesar Supplier No. 040129.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Silver hydrogen fluoride facilitates the photochemical conversion of carboxylic acids to alkyl fluorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12249-52-4
MDL Number MFCD00042145
Synonym silver hydron fluoride, silver salt, h, fluoride, silver 1+ ion hydrogen fluoride, hydrogen . silver 1+ fluoride

Specifications

Melting Point (decomposition)
Physical Form Crystalline
Quantity 10 g
Linear Formula AgHF2
UN Number UN1740
Solubility Information Soluble in water.
Formula Weight 146.87
Percent Purity ≤99%
Sensitivity Hygroscopic; Light sensitive
Chemical Name or Material Silver hydrogen fluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.