CAS RN 12249-52-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Silver hydrogen fluoride, 99% min, Thermo Scientific Chemicals
CAS: 12249-52-4 MDL Number: MFCD00042145 Synonym: silver hydron fluoride,silver salt, h, fluoride,silver 1+ ion hydrogen fluoride,hydrogen . silver 1+ fluoride