Learn More
Bollé Safety Silium+ Platinum™ Smoke Safety Glasses
Product Code. 15588291 Shop All Bollé Safety Products
Product Code. 15588291 Supplier Bollé Safety Supplier No. SILPPSF
  • ONLINE EXCLUSIVE PRICE
    £125.67
    £104.67 / Pack of 10
    Save £21.00
    16% Off
Estimated Shipment: 12-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Wear a comfortable and ultra lightweight polycarbonate model for applications that require extended wear. The SILIUM+ SILPPSF combines style, ergonomics and performances. The permanent PLATINUM coating is applied to both surfaces, making them extremely scratch and fog resistant
  • Ultra lightweight frame for extended wear
  • Adjustable bridge
  • Non-slip bridge
  • Straight, highly flexible ergonomic temples
  • PLATINUM coating
  • All-round vision
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Mechanical risk - Low energy impact - F; Mechanical risk- High speed particles at extreme temperatures - T; Risks relating to radiations - UV Protection - EN 170 - (2-1.2, 2C-1.2); Solar radiations risks: EN 172
Lens Coating Anti-fog, Scratch-resistant
Description Tinted
Lens Tint Smoke
Frame Color Black
Weight (Metric) 21 g
Lens Material Polycarbonate