Learn More
Hirschmann™ SILICOSEN™ Pink Silicone Stoppers
Excellent for the cultivation of aerobic microorganisms. Hirschmann Instruments™ SILICOSEN™ Silicone Stoppers are specially processed culture plugs with continuous bubbles for preparation, filling and sterilization of culture media.
Brand: Hirschmann™ 8905542
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Uniform pore structure
- Good air permeability
- Chemically and temperature resistant
- Liquid repellent and reusable
Specifications
Pink | |
30-40 mm | |
10/Pk. | |
35 mm |
Stopper | |
For The Preparation And Sterilization Of Culture Media | |
Silicone |