Learn More
Macherey-Nagel™ SILCEL-Mix-25 cellulose and silica mixed layer on glass plates
Product Code. 10756712 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensions (L x W):
20 x 20 cm
Unit Size:
Pack of 25
Product Code. Dimensions (L x W) Unit Size
10756712 20 x 20 cm Pack of 25
1 options
Product Code. 10756712 Supplier Macherey-Nagel™ Supplier No. 810043
  • ONLINE EXCLUSIVE PRICE
    £250.00
    £231.08 / Pack of 25
    Save £18.92
    7% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Separate preservatives and other antimicrobial compounds. SILCEL-Mix-25 Cellulose and Silica Mixed Layer on Glass Plates is the perfect choice in TLC separations.
  • Mixed layer of cellulose and silica
  • Recommended for separation of preservatives
  • Recommended for separation of antimicrobial compounds
Specifications
Material Cellulose and Silica
Quantity 25 Pk.
Type Glass Plate
Fluorescent Indicator UV254
Thickness (Metric) 0.25 mm
Dimensions (L x W) 20 x 20 cm