missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Silica Chromabond Diamino Adsorbent

Product Code. 12834431 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
20 g
100 g
Unit Size:
100g
20g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12834431 20 g 20g
10743324 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12834431 Supplier Macherey-Nagel™ Supplier No. 730653.20

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 20 g
Product Type Chromabond Diamino Adsorbent
Particle Size 45 μm
Material Silica
pH Range 2 to 8
Pore Size 60 Å
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.