Description
- Perfectly molded polypropylene tips for consistent accuracy of sampled volumes
- Sterile tips in blister pack
- Autoclavable and reusable
Specifications
Specifications
| Color | Blue |
| Autoclavable | Autoclavable |
| Product Line | Silex II |
| Material | Polypropylene |