missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIK1/Snf1lk Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38397
This item is not returnable.
View return policy
Description
SIK1/Snf1lk Polyclonal specifically detects SIK1/Snf1lk in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SIK1/Snf1lk | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| P57059 | |
| SIK1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: YLLLERLKEYRNAQCARPGPARQPRPRSSDLSGLEVPQEGLSTDPFRPALLCPQPQTLVQSVLQAEMDCELQSSLQWPLFFPVDASCSGVF | |
| 0.1 mL | |
| Protein Kinase | |
| 150094 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.1, msk, myocardial SNF1-like kinase, salt-inducible kinase 1, Salt-inducible protein kinase 1, serine/threonine protein kinase, serine/threonine-protein kinase SIK1, Serine/threonine-protein kinase SNF1-like kinase 1, Serine/threonine-protein kinase SNF1LK, SIK, SIK-1, SNF1-like kinase, SNF1LKMSK | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction