missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIGLEC9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | SIGLEC9 |
|---|---|
| Concentration | 0.36 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16841554
|
Proteintech
13377-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16831554
|
Proteintech
13377-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Siglec9 (Sialic acid-binding Ig-like lectin 9) is single-pass type I membrane protein and belongs to immunoglobulin super family and contains two Ig-like C2 type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. It is reported that Siglec9 recognizes sialic acid in alpha2, 3-or alpha2, 6-glycosidic linkage to galactose. Using a specific antibody, Siglec9 was found to be expressed at high or intermediate levels by monocytes, neutrophils, and a minor population of CD16+, CD56- cells. Siglec9 also acts as an inhibitory receptor for MUC16 that attenuates T- andNK cell function. Recent reports suggest that Siglec9 expressed on DCs is involved in immuno regulation through ligation with mucins in an epithelial cancer patient.Specifications
| SIGLEC9 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q9Y336 | |
| 27180, 83382 | |
| SIGLEC9 Fusion Protein Ag4201 | |
| Primary | |
| -20°C | |
| SIGLEC9 |
| 0.36 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CD329, CDw329, OBBP LIKE, Protein FOAP 9, Siglec 9, SIGLEC9 | |
| SIGLEC9, Siglece | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title