missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-7/CD328 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59244
This item is not returnable.
View return policy
Description
Siglec-7/CD328 Polyclonal specifically detects Siglec-7/CD328 in Human samples. It is validated for Western Blot.
Specifications
| Siglec-7/CD328 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec-7 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Bovine: 84%; Chimpanzee: 84%; Human: 100%;. | |
| Human, Bovine, Canine, Equine, Guinea Pig | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9Y286 | |
| SIGLEC7 | |
| Synthetic peptides corresponding to SIGLEC7(sialic acid binding Ig-like lectin 7) The peptide sequence was selected from the middle region of SIGLEC7. Peptide sequence WRSLTLYPSQPSNPLVLELQVHLGDEGEFTCRAQNSLGSQHVSLNLSLQQ. | |
| 100 μL | |
| Cardiovascular Biology | |
| 27036 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction