missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-3/CD33 Antibody (CD33 3D6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£159.00 - £302.00
Specifications
| Antigen | Siglec-3/CD33 |
|---|---|
| Clone | CD33 3D6 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100 - 1:2000 |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18655016
|
Novus Biologicals
NBP2-50291-0.025ml |
0.025 mL |
£159.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611148
|
Novus Biologicals
NBP2-50291 |
0.1 mL |
£302.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human samples. It is validated for ELISASpecifications
| Siglec-3/CD33 | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| PBS | |
| 945 | |
| IgG1 | |
| Protein A purified |
| CD33 3D6 | |
| ELISA 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67 | |
| Siglec-3/CD33-Fc recombinant protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title