missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Siglec-3/CD33 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | CD33 3D6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:100 - 1:2000 |
| Formulation | PBS |
| Gene Alias | CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?