Learn More
Kinematica™ Stainless Steel Sieve for Polymix™ MFC 90 D Mill
Product Code. 10134133 Shop All Kinematica Products
Change view
Click to view available options
Pore Size:
0.2 mm
0.5 mm
0.8 mm
1 mm
1.5 mm
2.0 mm
3 mm
4 mm
5 mm
6 mm
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
10134133 1.5 mm Each
10012393 0.8 mm Each
10387183 3 mm Each
10713454 5 mm Each
10754024 6 mm Each
10042293 0.2 mm Each
10275893 0.5 mm Each
10532274 1 mm Each
10617693 2.0 mm Each
10677123 4 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 missing translation for 'options'
Product Code. 10134133 missing translation for 'mfr' Kinematica™ missing translation for 'supplierNo' 35095015
Only null left
Add to basket
Add to basket
Depending on the sample, even finer results can be achieved by grinding with two or more sieves with progressively smaller hole sizes.
Specifications
Product Type Sieve
Material Stainless Steel
Pore Size 1.5 mm