missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHP-2/PTPN11 Antibody (6D9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00 - £467.00
Specifications
| Antigen | SHP-2/PTPN11 |
|---|---|
| Clone | 6D9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18162699
|
Novus Biologicals
NBP2-37454 |
0.1 mL |
£467.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673326
|
Novus Biologicals
NBP2-37454-0.025ml |
0.025 mL |
£196.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SHP-2/PTPN11 Monoclonal specifically detects SHP-2/PTPN11 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| SHP-2/PTPN11 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP-1D, PTP-2C, PTP2CNS1, Shp2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11 | |
| PTPN11 | |
| IgG1 | |
| Affinity Purified |
| 6D9 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Angiogenesis, Cancer, Cell Biology, Lipid and Metabolism, Signal Transduction, Tyrosine Kinases | |
| PBS with 0.05% Sodium Azide | |
| 5781 | |
| Purified recombinant fragment of human PTPN11 (AA: 263-329) expressed in E. Coli. | |
| Primary | |
| 68.4 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title