missing translation for 'onlineSavingsMsg'
Learn More

SHP-2/PTPN11 Antibody (6D9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-37454-0.025ml

 View more versions of this product

Product Code. 18673326

  • £196.00 / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SHP-2/PTPN11 Monoclonal specifically detects SHP-2/PTPN11 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SHP-2/PTPN11
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin, CyTOF-ready
BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP-1D, PTP-2C, PTP2CNS1, Shp2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11
Mouse
68.4 kDa
0.025 mL
Angiogenesis, Cancer, Cell Biology, Lipid and Metabolism, Signal Transduction, Tyrosine Kinases
5781
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
6D9
Unconjugated
PBS with 0.05% Sodium Azide
PTPN11
Purified recombinant fragment of human PTPN11 (AA: 263-329) expressed in E. Coli.
Affinity Purified
RUO
Primary
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only