missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHP-1 Mouse anti-Human, Clone: CPTC-PTPN6-2, Novus Biologicals™
Mouse Monoclonal Antibody
£215.00
Specifications
| Antigen | SHP-1 |
|---|---|
| Clone | CPTC-PTPN6-2 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, ELISA |
| Applications | Western Blot, ELISA |
Description
SHP-1 Monoclonal specifically detects SHP-1 in Human samples. It is validated for Western Blot, ELISA.Specifications
| SHP-1 | |
| 0.2 mg/ml | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Phospho Specific, Protein Phosphatase | |
| 10 mM PBS with 0.05% BSA | |
| 5777 | |
| IgG2a κ | |
| Protein A or G purified |
| CPTC-PTPN6-2 | |
| Western Blot 1-2 ug/ml, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| HCP, HCPH, HPTP1C, PTP-1C, PTPN6, SHP1, SHP-1L, SH-PTP1 | |
| Recombinant human full-length SHP-1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title