missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHP-1 Mouse anti-Human, Clone: CPTC-PTPN6-2, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07158-20UG
This item is not returnable.
View return policy
Description
SHP-1 Monoclonal specifically detects SHP-1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| SHP-1 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 ug/ml, ELISA | |
| HCP, HCPH, HPTP1C, PTP-1C, PTPN6, SHP1, SHP-1L, SH-PTP1 | |
| Recombinant human full-length SHP-1 protein | |
| 20 μg | |
| Apoptosis, Phospho Specific, Protein Phosphatase | |
| 5777 | |
| Store at 4°C. | |
| IgG2a κ |
| Western Blot, ELISA | |
| CPTC-PTPN6-2 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction