missing translation for 'onlineSavingsMsg'
Learn More

SHOX2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£159.00 - £366.00

Specifications

Antigen SHOX2
Dilution Western Blot 1:1000 - 1:5000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18614422
View Documents
Novus Biologicals
NBP2-94905-0.02ml
0.02 mL
£159.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18634142
View Documents
Novus Biologicals
NBP2-94905-0.1ml
0.1 mL
£366.00
0.01mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SHOX2 Polyclonal antibody specifically detects SHOX2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

SHOX2
Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
Homeobox protein Og12X, OG12SHOX homologous gene on chromosome 3, OG12Xshort stature homeobox protein 2, short stature homeobox 2, SHOTPaired-related homeobox protein SHOT
Recombinant fusion protein containing a sequence corresponding to amino acids 96-355 of human SHOX2 (NP_003021.3). ELDMGAAERSREPGSPRLTEGRRKPTKAEVQATLLLPGEAFRFLVSPELKDRKEDAKGMEDEGQTKIKQRRSRTNFTLEQLNELERLFDETHYPDAFMREELSQRLGLSEARVQVWFQNRRAKCRKQENQLHKGVLIGAASQFEACRVAPYVNVGALRMPFQQDSHCNVTPLSFQVQAQLQLDSAVAHAHHHLHPHLAAHAPYMMFPAPPFGLPLATLAADSASAASVVAAAAAAKTTSKNSSIADLRLKAKKHAAALGL
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:1000 - 1:5000
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
6474
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.