missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHOX2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94905-0.1ml
This item is not returnable.
View return policy
Description
SHOX2 Polyclonal antibody specifically detects SHOX2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SHOX2 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000 | |
| Homeobox protein Og12X, OG12SHOX homologous gene on chromosome 3, OG12Xshort stature homeobox protein 2, short stature homeobox 2, SHOTPaired-related homeobox protein SHOT | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 96-355 of human SHOX2 (NP_003021.3). ELDMGAAERSREPGSPRLTEGRRKPTKAEVQATLLLPGEAFRFLVSPELKDRKEDAKGMEDEGQTKIKQRRSRTNFTLEQLNELERLFDETHYPDAFMREELSQRLGLSEARVQVWFQNRRAKCRKQENQLHKGVLIGAASQFEACRVAPYVNVGALRMPFQQDSHCNVTPLSFQVQAQLQLDSAVAHAHHHLHPHLAAHAPYMMFPAPPFGLPLATLAADSASAASVVAAAAAAKTTSKNSSIADLRLKAKKHAAALGL | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6474 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction