missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79656
This item is not returnable.
View return policy
Description
SHC3 Polyclonal specifically detects SHC3 in Mouse samples. It is validated for Western Blot.
Specifications
| SHC3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Neuronal Shc, NSHCDKFZp686H1544, N-Shcsrc homology 2 domain containing transforming protein C3, Protein Rai, RAI, SH2 domain protein C3, SHC (Src homology 2 domain containing) transforming protein 3, SHCCFLJ45325, SHC-transforming protein 3, SHC-transforming protein C, src homology 2 domain-containing transforming protein C3, Src homology 2 domain-containing-transforming protein C3 | |
| Rabbit | |
| 52 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_033193 | |
| SHC3 | |
| Synthetic peptide directed towards the N terminal of human Shc3The immunogen for this antibody is Shc3. Peptide sequence PSKMLSSILGKSNLQFAGMSISLTISTASLNLRTPDSKQIIANHHMRSIS. | |
| Affinity purified | |
| RUO | |
| 53358 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction