missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHC1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00 - £348.00
Specifications
| Antigen | SHC1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18305283
|
Novus Biologicals
NBP3-16938-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18317965
|
Novus Biologicals
NBP3-16938-100UL |
100 μg |
£348.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SHC1 Polyclonal antibody specifically detects SHC1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| SHC1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| FLJ26504, p66, p66SHC, SH2 domain protein C1, SHC, SHC (Src homology 2 domain containing) transforming protein 1, SHC (Src homology 2 domain-containing) transforming protein 1, SHCA, SHC-transforming protein 1, SHC-transforming protein 3, SHC-transforming protein A, Src homology 2 domain-containing-transforming protein C1 | |
| A synthetic phosphorylated peptide around Y427 of human SHC1 (NP_892113.4). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Phospho Specific, Signal Transduction | |
| PBS, 50% glycerol, pH7.3 | |
| 6464 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title