missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHC1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-16938-100UL
This item is not returnable.
View return policy
Description
SHC1 Polyclonal antibody specifically detects SHC1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| SHC1 | |
| Polyclonal | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| FLJ26504, p66, p66SHC, SH2 domain protein C1, SHC, SHC (Src homology 2 domain containing) transforming protein 1, SHC (Src homology 2 domain-containing) transforming protein 1, SHCA, SHC-transforming protein 1, SHC-transforming protein 3, SHC-transforming protein A, Src homology 2 domain-containing-transforming protein C1 | |
| A synthetic phosphorylated peptide around Y427 of human SHC1 (NP_892113.4). | |
| 100 μg | |
| Cell Cycle and Replication, Phospho Specific, Signal Transduction | |
| 6464 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction